Condenser clamp
Catalog # JUCHVL04685
Supplier: JUCHHEIM Laborgerate GmbH
Specifications
- Larghezza di serraggio:85 mm
- Descrizione:Condenser clamp
- Environmentally Preferable:
- Conf:1
Specifications
About this item
Malleable iron, epoxy coated.
- With epoxy coating
- Movable support rod
Weight: 480 g