Counting chamber, Fast Read® 102
Catalog # 630-1893
Supplier: Kova International
Specifications
- L×P:5×2 mm
- Descrizione:Counting chambers with 10 squares and 16 grids each
- Conf:100
Specifications
About this item
Fast Read 102 systems are made of a disposable plastic slide and designed for the standardisation of microscopic urinalysis.
- Protected by an optically transparent film
- Precision and reproducibility guarantee constant readings
- Identification of the sample by using the numbers printed on the sides
Each chamber is fitted with its own system for the collection of excess urine to prevent any possible contamination.
Chamber depth: 0,1 mm
Chamber volume: 7 µl