Orders LinkContactUsLinkComponent
Human Recombinant G-CSF (from E. coli) | Avantor
Human Recombinant G-CSF (from E. coli)
Nome del fornitore:   Shenandoah Biotechnology
Human Recombinant G-CSF (from <i>E. coli</i>)
Nome del fornitore:  Shenandoah Biotechnology
Certificates
Human Recombinant G-CSF (from <i>E. coli</i>)
3 varianti
Product Details & Documents
Granulocyte-colony stimulating factor (G-CSF) is a cytokine that functions as a potent inducer of neutrophilic granulocyte proliferation, terminal differentiation, and activation. G-CSF synthesis occurs in monocyte, macrophage, epithelial, endothelial, and fibroblast cells after activation by bacterial endotoxins, tumor necrosis factor alpha (TNF-α), interleukin 1 (IL-1), or interleukin 17 (IL-17).
  • High quality
  • Low endotoxin
  • Affordable
The functional activity of G-CSF is mediated through the granulocyte colony-stimulating factor receptor (G-CSF-R) to activate JAK/STAT and MAPK signal transduction pathways.  G-CSF also promotes neurogenesis and inhibits neuronal apoptosis.  Human and mouse G-CSF proteins are cross-reactive.
The functional activity of G-CSF is mediated through the granulocyte colony-stimulating factor receptor (G-CSF-R) to activate JAK/STAT and MAPK signal transduction pathways. G-CSF also promotes neurogenesis and inhibits neuronal apoptosis. Human and mouse G-CSF proteins are cross-reactive.
Lieferinformationen: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Achtung! Bitte beachten: For research use only.
Specifications for Product Family
Specifiche
Specifiche
  • Catalog No:
  • SHBT100-72-10UG
  • SHBT100-72-100UG
  • SHBT100-72-1MG
  • Protein/peptide name:
  • G-CSF
  • G-CSF
  • G-CSF
  • Protein synonyms:
  • GM-CSFβ|pluripoietin|MGI-1G|CSF-3
  • GM-CSFβ|pluripoietin|MGI-1G|CSF-3
  • GM-CSFβ|pluripoietin|MGI-1G|CSF-3
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Specie:
  • Human
  • Human
  • Human
  • Sorgente luminosa:
  • E. coli
  • E. coli
  • E. coli
  • Coniugazione:
  • Non coniugato
  • Non coniugato
  • Non coniugato
  • ID gene:
  • P09919
  • P09919
  • P09919
  • Sequenza:
  • MTPLGPASSL PQSFLLKCLE QVRKIQGDGA ALQEKLCATY KLCHPEELVL LGHSLGIPWA PLSSCPSQAL QLAGCLSQLH SGLFLYQGLL QALEGISPEL GPTLDTLQLD VADFATTIWQ QMEELGMAPA LQPTQGAMPA FASAFQRRAG GVLVASHLQS FLEVSYRVLR HLAQP
  • MTPLGPASSL PQSFLLKCLE QVRKIQGDGA ALQEKLCATY KLCHPEELVL LGHSLGIPWA PLSSCPSQAL QLAGCLSQLH SGLFLYQGLL QALEGISPEL GPTLDTLQLD VADFATTIWQ QMEELGMAPA LQPTQGAMPA FASAFQRRAG GVLVASHLQS FLEVSYRVLR HLAQP
  • MTPLGPASSL PQSFLLKCLE QVRKIQGDGA ALQEKLCATY KLCHPEELVL LGHSLGIPWA PLSSCPSQAL QLAGCLSQLH SGLFLYQGLL QALEGISPEL GPTLDTLQLD VADFATTIWQ QMEELGMAPA LQPTQGAMPA FASAFQRRAG GVLVASHLQS FLEVSYRVLR HLAQP
  • Protease-free:
  • N
  • N
  • N
  • Senza animali:
  • N
  • N
  • N
  • Esente da nucleasi:
  • N
  • N
  • N
  • Senza corriere:
  • Y
  • Y
  • Y
  • Privo di endotossine:
  • N
  • N
  • N
  • Livello di endotossine:
  • Low
  • Low
  • Low
  • Contenuto di endotossine:
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • UniProtKB:
  • P09919
  • P09919
  • P09919
  • Attività biologica:
  • Bioactivity assay: NFS-60 cell proliferation, ED50 ≤0,060 ng/ml, bioactivity ≥1,7×10⁷ units/mg
  • Bioactivity assay: NFS-60 cell proliferation, ED50 ≤0,060 ng/ml, bioactivity ≥1,7×10⁷ units/mg
  • Bioactivity assay: NFS-60 cell proliferation, ED50 ≤0,060 ng/ml, bioactivity ≥1,7×10⁷ units/mg
  • Purezza:
  • ≥95%
  • ≥95%
  • ≥95%
  • Grado:
  • RUO
  • RUO
  • RUO
  • Formulazione:
  • 20 mM acetic acid, 50 mM sodium chloride
  • 20 mM acetic acid, 50 mM sodium chloride
  • 20 mM acetic acid, 50 mM sodium chloride
  • Peso molecolare:
  • 18,8 kDa
  • 18,8 kDa
  • 18,8 kDa
  • Ricostituzione:
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Sterile water at 0,1 mg/ml
  • Condizioni di conservazione:
  • −20 °C
  • −20 °C
  • −20 °C
  • Temperatura di spedizione:
  • RT
  • RT
  • RT
  • Applicazioni testate:
  • Bioassay
  • Bioassay
  • Bioassay
Specifiche
Product Family Options
Informazioni sul prodotto
  • Tipo di conf.
    Conf
    Disponibilità
    Price
  • Vial
    10 µG
    Log in per disponibilità
    Log in per disponibilità
    1 * 10 µG
    1 * 10 µG - 182.00 SFr.
    182.00 SFr.
    1 * 10 µG
       182.00 SFr.
    Specifiche:
    More
    Specifications
    Cat. No.
    SHBT100-72-10UG
    Protein/peptide name
    G-CSF
    Protein synonyms
    GM-CSFβ|pluripoietin|MGI-1G|CSF-3
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Specie
    Human
    Sorgente luminosa
    E. coli
    Coniugazione
    Non coniugato
    ID gene
    P09919
    Sequenza
    MTPLGPASSL PQSFLLKCLE QVRKIQGDGA ALQEKLCATY KLCHPEELVL LGHSLGIPWA PLSSCPSQAL QLAGCLSQLH SGLFLYQGLL QALEGISPEL GPTLDTLQLD VADFATTIWQ QMEELGMAPA LQPTQGAMPA FASAFQRRAG GVLVASHLQS FLEVSYRVLR HLAQP
    Protease-free
    N
    Senza animali
    N
    Esente da nucleasi
    N
    Senza corriere
    Y
    Privo di endotossine
    N
    Livello di endotossine
    Low
    Contenuto di endotossine
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    P09919
    Attività biologica
    Bioactivity assay: NFS-60 cell proliferation, ED50 ≤0,060 ng/ml, bioactivity ≥1,7×10⁷ units/mg
    Purezza
    ≥95%
    Grado
    RUO
    Formulazione
    20 mM acetic acid, 50 mM sodium chloride
    Peso molecolare
    18,8 kDa
    Ricostituzione
    Sterile water at 0,1 mg/ml
    Condizioni di conservazione
    −20 °C
    Temperatura di spedizione
    RT
    Applicazioni testate
    Bioassay
  • Vial
    100 µG
    Log in per disponibilità
    Log in per disponibilità
    1 * 100 µG
    1 * 100 µG - 587.00 SFr.
    587.00 SFr.
    1 * 100 µG
       587.00 SFr.
    Specifiche:
    More
    Specifications
    Cat. No.
    SHBT100-72-100UG
    Protein/peptide name
    G-CSF
    Protein synonyms
    GM-CSFβ|pluripoietin|MGI-1G|CSF-3
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Specie
    Human
    Sorgente luminosa
    E. coli
    Coniugazione
    Non coniugato
    ID gene
    P09919
    Sequenza
    MTPLGPASSL PQSFLLKCLE QVRKIQGDGA ALQEKLCATY KLCHPEELVL LGHSLGIPWA PLSSCPSQAL QLAGCLSQLH SGLFLYQGLL QALEGISPEL GPTLDTLQLD VADFATTIWQ QMEELGMAPA LQPTQGAMPA FASAFQRRAG GVLVASHLQS FLEVSYRVLR HLAQP
    Protease-free
    N
    Senza animali
    N
    Esente da nucleasi
    N
    Senza corriere
    Y
    Privo di endotossine
    N
    Livello di endotossine
    Low
    Contenuto di endotossine
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    P09919
    Attività biologica
    Bioactivity assay: NFS-60 cell proliferation, ED50 ≤0,060 ng/ml, bioactivity ≥1,7×10⁷ units/mg
    Purezza
    ≥95%
    Grado
    RUO
    Formulazione
    20 mM acetic acid, 50 mM sodium chloride
    Peso molecolare
    18,8 kDa
    Ricostituzione
    Sterile water at 0,1 mg/ml
    Condizioni di conservazione
    −20 °C
    Temperatura di spedizione
    RT
    Applicazioni testate
    Bioassay
  • Vial
    1 mg
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 mg
    1 * 1 mg - 4 240.00 SFr.
    4 240.00 SFr.
    1 * 1 mg
       4 240.00 SFr.
    Specifiche:
    More
    Specifications
    Cat. No.
    SHBT100-72-1MG
    Protein/peptide name
    G-CSF
    Protein synonyms
    GM-CSFβ|pluripoietin|MGI-1G|CSF-3
    Protein/peptide type
    Recombinant
    Protein function
    Cytokine
    Specie
    Human
    Sorgente luminosa
    E. coli
    Coniugazione
    Non coniugato
    ID gene
    P09919
    Sequenza
    MTPLGPASSL PQSFLLKCLE QVRKIQGDGA ALQEKLCATY KLCHPEELVL LGHSLGIPWA PLSSCPSQAL QLAGCLSQLH SGLFLYQGLL QALEGISPEL GPTLDTLQLD VADFATTIWQ QMEELGMAPA LQPTQGAMPA FASAFQRRAG GVLVASHLQS FLEVSYRVLR HLAQP
    Protease-free
    N
    Senza animali
    N
    Esente da nucleasi
    N
    Senza corriere
    Y
    Privo di endotossine
    N
    Livello di endotossine
    Low
    Contenuto di endotossine
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    P09919
    Attività biologica
    Bioactivity assay: NFS-60 cell proliferation, ED50 ≤0,060 ng/ml, bioactivity ≥1,7×10⁷ units/mg
    Purezza
    ≥95%
    Grado
    RUO
    Formulazione
    20 mM acetic acid, 50 mM sodium chloride
    Peso molecolare
    18,8 kDa
    Ricostituzione
    Sterile water at 0,1 mg/ml
    Condizioni di conservazione
    −20 °C
    Temperatura di spedizione
    RT
    Applicazioni testate
    Bioassay