Dimidio bromuro
Glass bottle for solids, 1 g
Catalog # MOLEM11783520
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 2,7-Diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-Diamino-5-methyl-6-phenylphenanthridinium bromide
- CAS Number:518-67-2
- Formula:C₂₀H₁₈BrN₃
- MDL Number:MFCD00011757
- Molecular Weight:380.29 g/mol
- Melting Pt:243…248 °C
- Storage Temperature:Ambiente
- EINECS:208-256-7
- UN:2811
- ADR:6.1,I
Specifications
Specifications
- Tipo di conf.:Glass bottle for solids
- Conf:1 g