Lutezio(III)ossido ≥99.9%
Catalog # 36244.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Conf:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: Lutetium oxide, Dilutetium trioxide
:
:397.93 g/mol
Melting Pt:2487 °C
EINECS:234-764-3
:GHS07
Storage Temperature:Ambiente
CAS Number:12032-20-1
Formula:Lu₂O₃
MDL Number:MFCD00011100
Density:9.420