Orders LinkContactUsLinkComponent
Ammonio solfato
 :   EUROCLONE
Ammonio solfato
 :  EUROCLONE
Ammonio solfato
 :  Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
  •  :7783-20-2
  •  :(NH₄)₂SO₄
  •  :MFCD00003391
  •  :132.14 g/mol
  •  :13,00559
  •  :336…339 °C
  •  :1,78 g/cm³ (20 °C)
  •  :Ambiente
  •  :231-984-1
  •  :01-2119455044-46
Product Family Options