Benzyltriethylammonium chloride
Glass bottle for solids, 100 g
Katalognr. MOLEM47480297
Marketsource
Anbieter: Molekula
Benzyltriethylammonium chloride
Katalognr. MOLEM47480297
Anbieter: Molekula
Marketsource
CAS Nummer: 56-37-1
Direkt zu
Product Details & Documents
Synonyme: Benzyl triethylammonium chloride
- CAS Nummer:56-37-1
- Formel:C₁₃H₂₂ClN
- MDL-Nummer:MFCD00011824
- Molekulargewicht:227.78 g/mol
- Schmelzender Platingehalt:190…192 °C
- Blitz Pt:>100 °C
- Lagertemperatur:Ambiente
- EINECS:200-270-1
Technische Daten
Technische Daten
- Tipo di conf.:Glass bottle for solids
- Conf:100 g