Worktop trolley, ESD
Worktop trolley, ESD, 750×600×655/1055 mm
Codice catalogo SOVEC70135001
Nome del fornitore: TRESTON
Vai a
Product Details & Documents
Worktop trolley for workspace with ESD-protection.
- Height adjustable from 655 to 1055 mm
- Four swivel wheels (Ø 100), two with brakes
Worktop and bottom shelf laminated ESD surface with beech edges.
Informazioni per l'ordine: Also available as non-ESD.
Specifiche
- Descrizione:Worktop trolley, ESD
- L×P×A:750×600×655/1055 mm