Order Entry
Switzerland
ContactUsLinkComponent
Sodio salicilato ≥99.5%, AnalaR NORMAPUR® per analisi
Irritant
Sodio salicilato ≥99.5%, AnalaR NORMAPUR® per analisi
Catalog # 28065.230
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Sodio salicilato ≥99.5%, AnalaR NORMAPUR® per analisi
Catalog # 28065.230
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Tipo di conf.:
    Bottiglia di plastica
  • Conf:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay (calculated on dried substance):
    Min. 99.5 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Water:
    Max. 0.2 %
  • Cl (Chloride):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 0.02 %
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

About this item

 :
 :160.10 g/mol
Merck Index:12,08819
Melting Pt:200 °C
Density:0.32 g/cm³ (20 °C)
EINECS:200-198-0
 :GHS07
Storage Temperature:Ambiente
CAS Number:54-21-7
Formula:HOC₆H₄COONa
MDL Number:MFCD00002440