Bis(2-idrossetil)-(iminotris)(idrossi metil)metano (BIS-TRIS)
Supplier: Roth Carl
Certificates
About this item
CAS Number:6976-37-0
:
Formula:C₈H₁₉NO₅
MDL Number:MFCD00002853
:209.24 g/mol
Melting Pt:102…103 °C
Storage Temperature:Ambiente
EINECS:230-237-7
Frequently Bought Together