Circulating cooling thermostats, unichillers, with Pilot ONE E-grade "Basic" controller
Informazioni su questo articolo
Unichillers offer better efficiency, stable pressure and delivery performance with a constant cooling water temperature compared to traditional tap water cooling. Chillers also reduce water consumption and help conserve the environment whilst lowering operating costs.
- Simple operation with menu navigation
- Robust stainless construction
- Easy to fill and empty
- Illuminated filling level indicator
- Bright, large OLED display
Unichillers are ideal for removing heat from chemical processes, cooling technical plants or as a central cooling water supply for laboratories. Unichillers are designed for continual operation at ambient temperatures up to 40 °C.
Pilot-ONE E-grade 'Basic' has wide-ranging functionality for classic temperature-control applications in the basic version. It allows the functionality to be extended at any time to suit new process requirements.
The Pilot ONE® controller is interchangeable and can be used as a remote control (with an optional data cable). It offers a brilliant 5.7" touchscreen with graphic functions and a comfortable navigation menu in 11 languages (German, English, French, Italian, Spanish, Portuguese, Czech, Polish, Russian, Chinese and Japanese) and USB and ethernet connections. User can set visual and acoustic alarms, limiting the temperature range, and program the unit with its clock and calendar functions for auto start for unsupervised startup. Powerful pressure pump, bench models with max. pump pressure of 3 bar have a built-in, adjustable bypass and a pressure gauge; in floor-standing models pressure control is carried out via external bypass (optional).
The Unichiller P models are equipped with high pressure pumps for applications with high pressure drops. T denotes tower models, w denotes water cooled models, wo denotes winter option.
The tower models offer power with a small footprint. These models are fitted with the new exchangeable Pilot ONE® controller and are used in both research and production.
Certifications: Comply with IP Class 20, Safety class III/FL.
Ordering information: Installation in an outside area is possible with winter operation packages. Please contact Avantor for details on other models or options and accessories available.
21 varianti
Product Details & Documents
Unichillers offer better efficiency, stable pressure and delivery performance with a constant cooling water temperature compared to traditional tap water cooling. Chillers also reduce water consumption and help conserve the environment whilst lowering operating costs.
- Simple operation with menu navigation
- Robust stainless construction
- Easy to fill and empty
- Illuminated filling level indicator
- Bright, large OLED display
Unichillers are ideal for removing heat from chemical processes, cooling technical plants or as a central cooling water supply for laboratories. Unichillers are designed for continual operation at ambient temperatures up to 40 °C.
Pilot-ONE E-grade 'Basic' has wide-ranging functionality for classic temperature-control applications in the basic version. It allows the functionality to be extended at any time to suit new process requirements.
The Pilot ONE® controller is interchangeable and can be used as a remote control (with an optional data cable). It offers a brilliant 5.7" touchscreen with graphic functions and a comfortable navigation menu in 11 languages (German, English, French, Italian, Spanish, Portuguese, Czech, Polish, Russian, Chinese and Japanese) and USB and ethernet connections. User can set visual and acoustic alarms, limiting the temperature range, and program the unit with its clock and calendar functions for auto start for unsupervised startup. Powerful pressure pump, bench models with max. pump pressure of 3 bar have a built-in, adjustable bypass and a pressure gauge; in floor-standing models pressure control is carried out via external bypass (optional).
The Unichiller P models are equipped with high pressure pumps for applications with high pressure drops. T denotes tower models, w denotes water cooled models, wo denotes winter option.
The tower models offer power with a small footprint. These models are fitted with the new exchangeable Pilot ONE® controller and are used in both research and production.
Certifications: Comply with IP Class 20, Safety class III/FL.
Ordering information: Installation in an outside area is possible with winter operation packages. Please contact Avantor for details on other models or options and accessories available.
Product Family Options
Air cooled models
Informazioni sul prodotto
- TipoRange di temperaturaPotenza di riscaldamentoPotenza di raffredd.PompaL×P×ADisponibilitàPrice
- Unichiller 007−20...+40 °C—0,7; 0,55; 0,4; 0,2 kW29 l/min, 1000 mbar350×496×622 mmLog in per disponibilità1 * 1 ST
- Unichiller 050−20...+40 °C—5,0; 4,2; 3,0; 1,8; kW48 l/min, 3400 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller 050-H−20...+100 °C2 kW5,0; 5,0; 4,2; 3,0; 1,8; kW48 l/min, 3400 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller 050 WO−20...+40 °C—4,2; 5,0; kW48 l/min, 3400 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller 050-H WO−20...+100 °C2 kW4,2; 5,0; kW48 l/min, 3400 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller 075 WO−20...+40 °C—7,5; kW48 l/min, 3400 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller 075 WO−20...+40 °C—5,3; 7,5; kW130 l/min, 5700 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller 075-H WO−20...+100 °C2 kW6,1; 7,5; kW48 l/min, 3400 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller 075-H WO−20...+100 °C25,3; 7,5; kW130 l/min, 5700 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller P050−20...+40 °C—5,0; 3,4; 2,3; 1,2; kW130 l/min, 5700 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller P050-H−20...+100 °C2 kW5,0; 5,0; 3,4; 2,3; 1,2; kW130 l/min, 5700 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller P050 WO−20...+40 °C—3,6; 5,0 kW140 l/min, 5300 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller P050-H WO−20...+100 °C2 kW3,4; 5,0 kW130 l/min, 5700 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller P100 WO−20...+40 °C—7,8; 10; kW130 l/min, 5700 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller P100-H WO−20...+100 °C2 kW7,8; 10; kW130 l/min, 5700 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller100 WO−20...+40 °C—8,6; 10; kW48 l/min, 3400 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller100-H WO−20...+100 °C2 kW8,6; 10; kW48 l/min, 3400 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
Water cooled models
Informazioni sul prodotto
- TipoRange di temperaturaPotenza di riscaldamentoPotenza di raffredd.PompaL×P×ADisponibilitàPrice
- Unichiller 075w−20...+40 °C—7,5; 6,1; 4,0; 2,4; kW48 l/min, 3400 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller 075w-H−20...+100 °C2 kW7,5; 7,5; 6,1; 4,0; 2,4; kW48 l/min, 3400 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller P075w−20...+40 °C—7,5; 5,3; 3,3; 1,8; kW130 l/min, 5700 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST
- Unichiller P075w-H−20...+100 °C2 kW7,5; 7,5; 5,3; 3,3; 1,8; kW130 l/min, 5700 mbar740×1160×1050 mmLog in per disponibilità1 * 1 ST