Orders LinkContactUsLinkComponent
L(+)-Glutammina
L(+)-Glutammina
L(+)-Glutammina
Supplier:  PanReac AppliChem
Certificates
L(+)-Glutammina
2 Options Available Below
Product Details & Documents
Synonyms:  Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
  • CAS Number:56-85-9
  • Formula:C₅H₁₀N₂O₃
  • MDL Number:MFCD00008044
  • Molecular Weight:146.15 g/mol
  • Merck Index:13,04484
  • Boiling Pt:355 °C (1013 hPa)
  • Melting Pt:185…186 °C
  • Density:1,5326 g/cm³ (130 °C)
  • Storage Temperature:Ambiente
  • EINECS:200-292-1
Documents
Product Family Options
Product Information
  • Tipo di conf.
    Conf
    Availability
    Price
  • Bottiglia di vetro
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - SFr. 185.00
    SFr. 185.00
    1 * 250 g
       SFr. 185.00
  • Bottiglia di vetro
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - SFr. 641.00
    SFr. 641.00
    1 * 1 kg
       SFr. 641.00