VWR®, Blocking buffer, non-fat powdered milk
Catalog # M232-39.8G-5PK
Supplier: VWR Chemicals
CAS Number:
Specifications
- Conf:199 g
- Descrizione:Blocking buffer, PBS with non fat milk, powder blend
Specifications
About this item
Each pack of powdered blocking buffer makes one liter of 1X solution.
- Easy to use
- Optimise assay performance using high quality blocking buffers