pH/mV/°C meters, handheld, Eutech pH 5+ / pH 6+
Anbieter: EUTECH
Zertifikate
Über diesen Artikel
These portable meters either measure pH and temperature (pH 5+) or pH, temperature and ORP values (pH 6+). With their compact and sturdy design, the lightweight meters are the ideal choice for field and lab work.
- Multi-point calibration (up to 5 points) with auto-buffer recognition - choice of USA, NIST or pure water buffer standards with ±0,01 pH accuracy
- Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
- Self-diagnostic messages for easy trouble-shooting
- Shock-resistant and splash resistant ABS plastic casing, rubber boot protects against knocks and hits
- HOLD function, automatic power-off after 20 minutes of inactivity
- Convenient benchtop stand, tactile keypad and customised LCD screen
Typical applications include: Pools and spas, ponds and aquariums, agriculture and hydroponics, water and wastewater treatment, cooling towers, food processing, photo-development and printing, ecological studies, field, laboratory and education institutions.
Technische Daten
- Catalog No:
- EUTEECPH601PLUSK
- EUTEECPH503PLUSK
- EUTEECPH501PLUS
- 662-2287
- EUTEECPH502PLUSK
- EUTEECPH601PLUS
- 662-2286
- Modello:
- Eutech pH 6+
- Eutech pH 5+
- Eutech pH 5+
- Eutech pH 6+
- Eutech pH 5+
- Eutech pH 6+
- Eutech pH 6+
- pH - range:
- 0,00 - 14,00
- 0,00 - 14,00
- 0,00 - 14,00
- 0,00 - 14,00
- 0,00 - 14,00
- 0,00 - 14,00
- 0,00 - 14,00
- pH - risoluzione:
- 0,01
- 0,01
- 0,01
- 0,01
- 0,01
- 0,01
- 0,01
- pH - accuratezza:
- ±0,01
- ±0,01
- ±0,01
- ±0,01
- ±0,01
- ±0,01
- ±0,01
- pH - calibrazione:
- Up to 5 points
- Up to 5 points
- Up to 5 points
- Up to 5 points
- Up to 5 points
- Up to 5 points
- Up to 5 points
- mV - range:
- ±1000
- -
- -
- ±1000
- -
- ±1000
- ±1000
- mV - risoluzione:
- 1
- -
- -
- 1
- -
- 1
- 1
- mV - accuratezza:
- ±2
- -
- -
- ±2
- -
- ±2
- ±2
- Range di temperatura:
- 0,0...+100,0 °C
- 0,0...+100,0 °C
- 0,0...+100,0 °C
- 0,0...+100,0 °C
- 0,0...+100,0 °C
- 0,0...+100,0 °C
- 0,0...+100,0 °C
- Risoluzione della temperatura:
- 0,1 °C
- 0,1 °C
- 0,1 °C
- 0,1 °C
- 0,1 °C
- 0,1 °C
- 0,1 °C
- Accuratezza della temperatura:
- ±0,5 °C
- ±0,5 °C
- ±0,5 °C
- ±0,5 °C
- ±0,5 °C
- ±0,5 °C
- ±0,5 °C
- Temperatura - compensazione:
- ATC/MTC (0...+100 °C)
- ATC/MTC (0...+100 °C)
- ATC/MTC (0...+100 °C)
- ATC/MTC (0...+100 °C)
- ATC/MTC (0...+100 °C)
- ATC/MTC (0...+100 °C)
- ATC/MTC (0...+100 °C)
- Display:
- LCD
- LCD
- LCD
- LCD
- LCD
- LCD
- LCD
- Alimentazione:
- 4× 1,5 V ‘AAA’ alkaline batteries
- 4× 1,5 V ‘AAA’ alkaline batteries
- 4× 1,5 V ‘AAA’ alkaline batteries
- 4× 1,5 V ‘AAA’ alkaline batteries
- 4× 1,5 V ‘AAA’ alkaline batteries
- 4× 1,5 V ‘AAA’ alkaline batteries
- 4× 1,5 V ‘AAA’ alkaline batteries
- Conformità:
- IP54
- IP54
- IP54
- IP54
- IP54
- IP54
- IP54
- Interfacce:
- BNC, 2.5 mm phono socket
- BNC, 2.5 mm phono socket
- BNC, 2.5 mm phono socket
- BNC, 2.5 mm phono socket
- BNC, 2.5 mm phono socket
- BNC, 2.5 mm phono socket
- BNC, 2.5 mm phono socket
- L×P×A:
- 85×42×157 mm
- 85×42×157 mm
- 85×42×157 mm
- 85×42×157 mm
- 85×42×157 mm
- 85×42×157 mm
- 85×42×157 mm
- Peso:
- 0,26 kg
- 0,26 kg
- 0,26 kg
- 0,26 kg
- 0,26 kg
- 0,26 kg
- 0,26 kg
Technische Daten
Electrodes
Produktinformationen
Frequently Bought Together