IHC enhancer (DAB, plus), HIGHDEF®
HIGHDEF® IHC enhancer (DAB, plus), 1X, 15 mL
Codice catalogo ENZOADI9501820015
Nome del fornitore: Enzo Life Sciences
IHC enhancer (DAB, plus), HIGHDEF®
Codice catalogo ENZOADI9501820015
Nome del fornitore: Enzo Life Sciences
Numero CAS:
Vai a
Product Details & Documents
HIGHDEF® IHC enhancer (DAB, plus) is a DAB signal enhancer used to increase intensity of immunostaining using DAB as chromogen.
- Superior performance compared to amino ethyl carbazol (AEC)
HIGHDEF® IHC enhancer (DAB, plus) increases the signal intensity without creating any background staining.
Specifiche
Specifiche
- Conf:15 mL
- Descrizione:HIGHDEF® IHC enhancer (DAB, plus), 1X