Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Inserisci ordine
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Misuratori elettrochimici
Misuratori per ossigeno disciolto
Accessories for eutech DO 450
Accessories for eutech DO 450
Print
Accessories for eutech DO 450
Zero oxygen solution, 0 % DO, 480 ml
Catalog # EUTEECDOZEROSOLN
Certificates
Supplier:
EUTECH
Accessories for eutech DO 450
Catalog # EUTEECDOZEROSOLN
Supplier:
EUTECH
Certificates
Expand image
Return to Top
Accessories for eutech DO 450
Zero oxygen solution, 0 % DO, 480 ml
Specifications
Add to cart
Jump to:
Specifications
Price
SFr. 31.00
Log in
for availability
Unit of Measure
1 * 1 ST
-
+
Add to cart
Return Policy
Product Details & Documents
Documents
Certificates
Search by Lot Number
Specifications
Descrizione:
Zero oxygen solution, 0 % DO, 480 ml
VWR®, Crogioli, Porcellana
#: 459-0208
Supplier: VWR Collection
VWR® Traceable®, Termometro antiurto
#: 620-2065
Supplier: VWR Collection