Orders LinkContactUsLinkComponent
Tween® 80
Supplier:   EUROCLONE
Tween® 80
Supplier:  EUROCLONE
Certificates
3 Options Available Below
Product Details & Documents
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Merck Index:13,07664
  • Boiling Pt:∼100 °C (1013 hPa)
  • Melting Pt:–21 °C
  • Density:1,062 g/cm³ (20 °C)
  • Flash Pt:>149 °C
  • Storage Temperature:Ambiente
  • EINECS:500-019-9
Specifications for Product Family
Specifications
Product Family Options
Product Information
  • Conf
    Availability
    Price
  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
    More
    Specifications
    Cat. No.
    EURLEMR245001
  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
    More
    Specifications
    Cat. No.
    EURLEMR245005
  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
    More
    Specifications
    Cat. No.
    EURLEMR245025