Paraformaldeide, EMPROVE® ESSENTIAL DAC, SAFC®
Catalog # 1.04005.5000
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Specifications
- Tipo di conf.:Bottiglia di plastica
- Conf:5 kg
- Seq:4114
Specifications
About this item
:
Merck Index:13,07096
Boiling Pt:135…136 °C (13 mmHg)
Melting Pt:123 °C
Density:1.46 g/cm³ (20 °C)
Flash Pt:70 °C
UN:2213
ADR:4.1,III
:GHS05
:GHS08
:GHS07
Storage Temperature:Refrigerata
CAS Number:30525-89-4
Formula:(CH₂O)n
MDL Number:MFCD00133991