Polietilenglicole 2.000
Supplier: Roth Carl
Certificates
About this item
Synonyms: PEG 2000, Ethylene glycol polymer 2000, PEG 2,000, Ethylene glycol polymer average Mn 2000
CAS Number:25322-68-3
:
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839
Density:1.21 g/cm³ (20 °C)
Molecular Weight:2000 g/mol
Melting Pt:49…52 °C
Flash Pt:250
Storage Temperature:Ambiente
EINECS:500-038-2
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together