Orders LinkContactUsLinkComponent
Gelatin
Supplier:   EUROCLONE
Gelatin
Supplier:  EUROCLONE
Certificates
3 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • EURLEMR142500
  • EURLEMR142001
  • EURLEMR142000
  • Protein/peptide name:
  • Gelatin
  • Gelatin
  • Gelatin
  • Protein synonyms:
  • Gelatine
  • Gelatine
  • Gelatine
  • CAS No.:
  • 9000-70-8
  • 9000-70-8
  • 9000-70-8
  • Condizioni di conservazione:
  • Ambient
  • Ambient
  • Ambient
Specifications
Product Family Options
Product Information
  • Tipo di conf.
    Conf
    Availability
    Price
  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT
    Specifications:
    More
    Specifications
    Cat. No.
    EURLEMR142500
    Protein/peptide name
    Gelatin
    Protein synonyms
    Gelatine
    CAS No.
    9000-70-8
    Condizioni di conservazione
    Ambient
  • Bottiglia di vetro
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg
    Specifications:
    More
    Specifications
    Cat. No.
    EURLEMR142001
    Protein/peptide name
    Gelatin
    Protein synonyms
    Gelatine
    CAS No.
    9000-70-8
    Condizioni di conservazione
    Ambient
  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg
    Specifications:
    More
    Specifications
    Cat. No.
    EURLEMR142000
    Protein/peptide name
    Gelatin
    Protein synonyms
    Gelatine
    CAS No.
    9000-70-8
    Condizioni di conservazione
    Ambient