Dimetilsolfossido-D6, dehydrated (max. 0,02% H₂O) (99.8% D) per spettroscopia NMR
Supplier: EURISO TOP
Certificates
About this item
CAS Number:2206-27-1
:
Formula:D₃CS(O)CD₃
MDL Number:MFCD00002090
:84.09 g/mol
Boiling Pt:189 °C (5 mmHg)
Melting Pt:20.2 °C
Density:1.19 g/cm³ (20 °C)
Flash Pt:82 °C (closed cup)
Storage Temperature:Ambiente
EINECS:218-617-0
Frequently Bought Together