Wound cleansers, QuickClean
Catalog # PLUM5551
Supplier: Plum Safety
Specifications
- Descrizione:QuickClean wound cleanser refill box, 40 wipes
- Conf:40
Specifications
About this item
These moist wipes are ideal for the quick and easy cleansing of minor wounds and cuts.
- Wall-mounted wound cleanser box with 40 wipes
- Hygienic: Each wipe is separately packed
- QuickClean wipes contain sterile water
Wipes L×W: 130×200 mm