Eyewash station, Combibox DUO
Codice catalogo PLUM4862
Nome del fornitore: Plum Safety
Specifiche
- Descrizione:Eyewash station, Combibox DUO, incl. 500 ml pH Neutral DUO and 500 ml Plum eye wash DUO
- Conf:1
Specifiche
Informazioni su questo articolo
Wall-mounted eye wash box made from maintenance-free material for quick first aid of eye injuries. The box contains one 500 ml bottle of pH Neutral DUO and one 500 ml bottle of Plum eye wash DUO.
- Dust-tight wall station in high-impact PS
- Quick and efficient eye wash
- Quick and easy to mount on the wall close to the workplace
- With clear pictograms and mirror
- Particularly suited for dusty and dirty production areas
W×D×H: 230×110×280 mm