Argento 99.9% (trace metals basis), in filo, Ø 2.0 mm , 1ea = 16.5gr
ACRO317720010
:
- Tipo di conf.:Plastic bag
- Conf:1
- Seq:0001
- Environmentally Preferable:
- Appearance:Silver white Wire
- Total trace metal impurities:≤0.1 %
:
:107.87 g/mol
:12,08647
:2212 °C (1013 hPa)
:961 °C
:10,5 g/cm³ (20 °C)
:Ambiente
:231-131-3
:7440-22-4
:Ag
:MFCD00003397