Soil test kit, NPK
LAMO3-5880
:
- Conf:1
- Descrizione:NPK soil text kit
Kit is sufficient to perform up to fifty tests each for nitrogen, phosphorus, and potassium levels using non-hazardous TesTabs.
- Kit is fun to use and easy to ship anywhere
- Ideal for upper elementary, middle, and high school students
It is specifically designed to meet the needs of the GLOBE ProgramTM (Global Learning and Observations to Benefit the Environment).
: It includes the necessary apparatus, simple diagrammed instructions, and a laminated colour chart.