Heavy-Duty Bin
50 L, 330/420 mm x 550 mm
Catalog # LAMA47/50
Supplier: LAMAPLAST
Jump to:
Product Details & Documents
Made of HDPE, natural.
- Good chemical resistance
- Sturdy
- Easy to clean
Bin with two side handles is suitable for food contact.
Certifications: Meets FCM regulations 1935/2004/CE and UE 10/2013.
Specifications
- Ø×A:330/420 mm x 550 mm
- Capacità:50 L