Measuring cup, graduated
500 ml, 115 mm, 110 mm
Codice catalogo USBE4400
Nome del fornitore: USBECK Laborgeräte
Vai a
Product Details & Documents
Cup made of stainless steel, conical.
- Graduated
- With handle
- Good chemical and temperature resistance
Specifiche
- Ø est:110 mm
- Capacità:500 ml
- Altezza:115 mm