Particulate filter, P3 R
Particle filter, P3
Codice catalogo BART922404
Nome del fornitore: BARTELS RIEGER
Vai a
Product Details & Documents
The high-performance particle filter for filtering or purifying air has a high-quality housing made from plastic.
- Particle separation efficiency of >99,95%
- Low inhalation resistance
- Low weight and compact design
The particle filter is designed for use in areas contaminated by dusts and aerosols.
Certifications: EN 133, DIN EN 148-1, EN 143:200
Specifiche
- Tipo:Particle filter
- Classe:P3