Test kits, nitrite, Comparator 2000+
Codice catalogo LOVI502371
Nome del fornitore: Lovibond
Numero CAS:
Specifiche
- Metodo chimico:N-(1-Naphthyl)ethylenediamine
- Conf:250
- Descrizione:Nitrite acidifying tablet
- Range di misura:0,002 – 0,05 mg/l N
Specifiche
Informazioni su questo articolo
Comparator 2000+ is a versatile modular system for testing water.
- Stable and accurate
- Robust, compact and portable
- Diffuser plate is used to measure the turbidity
- Longer path length cells be used for lower concentration analysis
The integrated prism brings the glass standards of the test discs and the coloured sample into the same field of view.