Human thrombin (from plasma)
1 Vial
Codice catalogo ENZOBMLSE3631000
Nome del fornitore: Enzo Life Sciences
Human thrombin (from plasma)
Codice catalogo ENZOBMLSE3631000
Nome del fornitore: Enzo Life Sciences
Numero CAS:
Vai a
Product Details & Documents
Isolated from human plasma as prothrombin, and activated with Factor Xa, Factor Va, phospholipids, and calcium to yield a-thrombin.
Specifiche
Specifiche
- Conf:1 Vial
- Cat. No.:ENZOBMLSE3631000
- Nome dell'enzima:Thrombin
- Purezza:Purified
- Specie:Human
- Sorgente luminosa:Plasma
- Attività enzimatica:1000 U
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



