ContactUsLinkComponent
Human recombinant p70 S6 Kinase
Human recombinant p70 S6 Kinase
Catalog # ENZOBMLSE3450005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant p70 S6 Kinase
Catalog # ENZOBMLSE3450005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Conf:
    5 µG
  • Tipo di enzima:
    Recombinant
  • Specie:
    Human
  • Seq:
    02
  • Nome dell'enzima:
    p70 S6 Kinase
Specifications
About this item
Produced in insect cells. Active, full length p70 S6K.