Human recombinant FGF-basic 147
Produced in E. coli. Non-glycosylated protein, containing 147 amino acids.
- Carrier-free
Produced in E. coli. Non-glycosylated protein, containing 147 amino acids.
- Carrier-free
- Catalog No:
- ENZOALX2018220010
- ENZOALX2018220050
- Protein/peptide name:
- FGF-basic 147
- FGF-basic 147
- Protein synonyms:
- Prostatropin|HBGF-2|FGF2
- Prostatropin|HBGF-2|FGF2
- Protein/peptide type:
- Recombinant
- Recombinant
- Specie:
- Human
- Human
- Attività biologica:
- The activity is determined by the dose-dependent proliferation of mouse BALB/c 3T3 cells which is typically observed at concentations <1 ng/ml.
- The activity is determined by the dose-dependent proliferation of mouse BALB/c 3T3 cells which is typically observed at concentations <1 ng/ml.
- Purezza:
- >95% (Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
- >95% (Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
- Formulazione:
- Lyophilized from 10 mM Na2PO4, pH 8,0. Sterile filtered.
- Lyophilized from 10 mM Na2PO4, pH 8,0. Sterile filtered.
- Peso molecolare:
- ~16,5kDa
- ~16,5kDa
Product Family Options
- Conf
Specifications
Cat. No.ENZOALX2018220010Protein/peptide nameFGF-basic 147Protein synonymsProstatropin|HBGF-2|FGF2Protein/peptide typeRecombinantSpecieHumanAttività biologicaThe activity is determined by the dose-dependent proliferation of mouse BALB/c 3T3 cells which is typically observed at concentations <1 ng/ml.Purezza>95% (Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)FormulazioneLyophilized from 10 mM Na2PO4, pH 8,0. Sterile filtered.Peso molecolare~16,5kDaSpecifications
Cat. No.ENZOALX2018220050Protein/peptide nameFGF-basic 147Protein synonymsProstatropin|HBGF-2|FGF2Protein/peptide typeRecombinantSpecieHumanAttività biologicaThe activity is determined by the dose-dependent proliferation of mouse BALB/c 3T3 cells which is typically observed at concentations <1 ng/ml.Purezza>95% (Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)FormulazioneLyophilized from 10 mM Na2PO4, pH 8,0. Sterile filtered.Peso molecolare~16,5kDa