Human recombinant neprilysin (from insect cells (Sf9)), HIS tag
ENZOBMLSE5320010
:
- Conf:10 µG
- Protein/peptide type:Recombinant
- Sorgente luminosa:Insect cells (Sf9)
- Specie:Human
- Seq:02
- Sequenza tag:HIS Tag
- Protein/peptide name:Neprilysin
- Purezza:>95% (SDS-PAGE)
- Concentrazione:0,23 μg/μl
- Formulazione:Liquid. In 50 mM TRIS, pH 8,0. containing 300 mM sodium chloride and 20% glycerol.
- Purificazione:Purified as the glycosylated soluble form.
Highly active
Produced in Sf9 cells.
Neprilysin is a glycosylated zinc endopeptidase whose physiological targets include bradykinin, glucagon, GLP-1, and neuropeptides. Preferential cleavage of polypeptides between hydrophobic residues, particularly with Phe or Tyr at P1.