Ac-REQD-pNA
5 mg
Codice catalogo ENZOBMLP4420005
Nome del fornitore: Enzo Life Sciences
Vai a
Product Details & Documents
Caspase substrate
Colorimetric caspase substrate. Sequence corresponds to the prodomain of procaspase-8 cleavage site, residues 213-216, a putative caspase-10 target. Selective caspase-1 substrate at concentrations of 0,2 to 2 mM.
Specifiche
Specifiche
- Conf:5 mg
- Cat. No.:ENZOBMLP4420005
- Protein/peptide name:Ac-REQD-pNA
- Purezza:95%