Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant LIGHT (soluble)
Human recombinant LIGHT (soluble)
Codice catalogo ENZOALX522018C010
Nome del fornitore:  ENZO LIFE SCIENCES
Numero CAS:  
Human recombinant LIGHT (soluble)
Codice catalogo ENZOALX522018C010
Nome del fornitore:  ENZO LIFE SCIENCES
Numero CAS:  

Specifiche

  • Conf:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Specie:
    Human
  • Seq:
    02
  • Attività biologica:
    Increases LNCaP (prostate cancer cell) growth.
  • Ricostituzione:
    Reconstitute with 100 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum.
  • Protein/peptide name:
    LIGHT (soluble)
  • Purezza:
    >95% (SDS-PAGE)
  • Concentrazione:
    0,1 mg/ml after reconstitution
  • Formulazione:
    Lyophilised. Contains PBS.

Specifiche

Informazioni su questo articolo

Produced in HEK 293 cells. The extracellular domain of human LIGHT (aa 89-240) is fused at the N-terminus to a linker peptide (10 aa) and a FLAG®-tag.