IHC PBS tablets
ENZOADI9502400100
New Product
:
- Conf:100
- Descrizione:IHC PBS tablets
- Seq:02
- Environmentally Preferable:
Solid. 100 ml PBS (1X) tablets containing 137 mM sodium chloride, 2.7 mM potassium chloride, and 10 mM phosphate buffer.