Cathepsin S Assay Buffer
Catalog # ENZOBMLKI4310020
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Conf:20 mL
- Descrizione:Cathepsin S assay buffer
- Seq:02
- Environmentally Preferable:
Specifications
About this item
50mM potassium phosphate buffer, pH 6,5, 50 mM NaCl, 2 mM EDTA, 0,01% Triton X-100®, 0,5 mM DTT.