Orders LinkContactUsLinkComponent
AEC Peroxidase substrate kit
AEC peroxidase substrate kit, 300 mL
  ENZOENZ43825
AEC Peroxidase substrate kit
  ENZOENZ43825
 :  
AEC Peroxidase substrate kit
AEC peroxidase substrate kit, 300 mL
 :  
SFr. 238.00
1 * 300 mL
The AEC Peroxidase Substrate Kit contains reagents for visualization of peroxidase reactions using Aminoethylcarbazole (AEC). In the presence of H2O2, peroxidases catalyze the oxidation of AEC into a brick red precipitate. The supplied reagents are sufficient for preparation of 300 ml of ready-to-use color reaction mixture.
 : Contents: Aminoethylcarbazole, 6 ml 20 mg/ml in dimethylformamide 8X Reaction Buffer/Substrate Reagent, 40 ml Hydrogen peroxide in acetate buffer with stabilizers and enhancers Mixing Bottle, 1 15 ml bottle for preparation of color reaction mixture
  • Conf:
    300 mL
  • Descrizione:
    AEC peroxidase substrate kit