Orders LinkContactUsLinkComponent
PKA kinase activity kit
PKA kinase activity kit
PKA kinase activity kit, 1 KIT
Codice catalogo ENZOADIEKS390A
Nome del fornitore:   Enzo Life Sciences
PKA kinase activity kit
Codice catalogo ENZOADIEKS390A
Nome del fornitore:  Enzo Life Sciences
Numero CAS:  
PKA kinase activity kit
PKA kinase activity kit, 1 KIT
Numero CAS:  
Price
914.00 SFr.
Log in per disponibilità
1 * 1 KIT
Product Details & Documents
The PKA Kinase activity kit is an efficient assay that utilizes little sample volume. This kit gives you the ability to an end-point or kinetic assay read-out in a convenient 96-well plate based assay and offers easy sample handling protocols. The assay also offers a high signal to background ratio.
For the quantitative determination of PKA kinase activity in partially purified, purified, or crude enzyme preparations from any species.
Informazioni per l'ordine: Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2
Specifiche
Specifiche
  • Conf:
    1 KIT
  • Descrizione:
    PKA kinase activity kit
  • Cat. No.:
    ENZOADIEKS390A
  • Taglia:
    96 wells