Cell counting kit-F
Nome del fornitore: Enzo Life Sciences
Informazioni su questo articolo
Fluorometric kit for simple and accurate cell proliferation and cytotoxicity assays. Advantages includes no radioisotopes, more sensitive than the colorimetric assay, short staining reaction time, no solubilization step, correlates with the [3H]-thymidine incorporation assay.
2 varianti
Product Details & Documents
Fluorometric kit for simple and accurate cell proliferation and cytotoxicity assays. Advantages includes no radioisotopes, more sensitive than the colorimetric assay, short staining reaction time, no solubilization step, correlates with the [3H]-thymidine incorporation assay.
Specifications for Product Family
Specifiche
Specifiche
- Catalog No:
- ENZOALX850245KI01
- ENZOALX850245KI02
- Applicazione:
- Fluorescence Microscopy
- Fluorescence Microscopy
- Taglia:
- 500 tests
- 5×500 tests
- Condizioni di conservazione:
- −20 °C
- −20 °C
Specifiche
Product Family Options
Informazioni sul prodotto
- Test per kitConfDisponibilitàPrice
- ENZOALX850245KI01Cell counting kit-FSpecifiche:
More
Specifications
Cat. No.ENZOALX850245KI01ApplicazioneFluorescence MicroscopyTaglia500 testsCondizioni di conservazione−20 °C - ENZOALX850245KI02Cell counting kit-FSpecifiche:
More
Specifications
Cat. No.ENZOALX850245KI02ApplicazioneFluorescence MicroscopyTaglia5×500 testsCondizioni di conservazione−20 °C