Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Anti-MDA5 Rabbit Polyclonal Antibody

Anti-MDA5 Rabbit Polyclonal Antibody

Catalog # ENZOENZABS2990100
Supplier:   Enzo Life Sciences
Anti-MDA5 Rabbit Polyclonal Antibody
Catalog # ENZOENZABS2990100
Supplier:  Enzo Life Sciences
Anti-MDA5 Rabbit Polyclonal Antibody
Price
SFr. 585.00
Log in for availability
1 * 100 µG
Product Details & Documents

Recommended Applications: ELISA, IF, IHC (PS), WB

Species reactivity: Human, Mouse

Type: Primary
Antigen: MDA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Specifications
  • Seq:
    02
  • Western blot:
    Yes
  • Reattività:
    Human
  • Immunogeno:
    Synthetic peptide corresponding to 16 amino acids from near the center of human MDA5
  • Purificazione:
    Affinity purified
  • Tampone di conservazione:
    In PBS containing 0,02% sodium azide
  • Coniugazione:
    Non coniugato
  • Isotipo:
    IgG
  • Cat. No.:
    ENZOENZABS2990100
  • Sinomini antigene:
    RLR-2|Helicard|CADM-140 autoantigen|RIG-I-like receptor 2|RH116|Interferon-induced helicase C domain-containing protein 1
  • ImmunoFluorescenza:
    Yes
  • Forma:
    Liquid
  • Simbolo dell'antigene:
    MDA5
  • Clonalità:
    Policlonale
  • Ospite:
    Rabbit
  • Temperatura di spedizione:
    Shipped on blue ice
  • Tipo anticorpo:
    Primary
  • Temperatura di conservazione:
    –20 °C
  • Tipo di anticorpi:
    Melanoma differentiation-associated gene 5
  • ImmunoChimica:
    Yes
  • ELISA:
    Yes