Specifications
- Modello:Image Xpress Nano
- Descrizione:Imaging system with MetaXpress software
- Seq:001
- Conf:1
Specifications
About this item
The ImageXpress Nano enables researchers to get better data faster, with automated microscopy to scale-up experiments to multi-slide or microplate level evaluations.
- Capture high quality images and garner more relevant quantitative data
- Configure with your preferred filters and objectives
- Transmitted light option for imaging of non labelled samples
- Option for sample incubation - temperature, humidity, CO₂ - provided through our environmental control feature
Automated microscopy allows researchers to scale-up their experiments to multi-slide or microplate-level evaluations: The Image Xpress Nano features a long life, solid state, light engine and optics to reliably deliver the right assay sensitivity. The user friendly software interface is easy to learn, and provides the ability to remotely configure, run acquisition protocols, and review analysis results.
Combined with MetaXpress high content image acquisition and analysis software, the ImageXpress Nano is an end-to-end solution to interpret images, understand data and explore ideas.