ContactUsLinkComponent
Screw caps for culture tubes, KIMBLE®
  SCERSP73805B-15415
undefined
Screw caps for culture tubes, KIMBLE®
  SCERSP73805B-15415
  • Filettatura:
    15-415
  • Colore:
    naturale
  • Descrizione:
    With closed top, linerless
  • Materiale:
    PP
  • Conf:
    1.000
PP.
  • Very good chemical resistance
  • Very good temperature resistance