Foam insert, SecureFit™
Foam insert for 3M™ SecureFit™ safety spectacles 600 series
Codice catalogo 111-2291
Nome del fornitore: 3M
Vai a
Product Details & Documents
3M™ SecureFit Foam insert provides a close fit and fills the gap between the eyewear and the wearer’s face.
- Provides close and secure fit
- Reduces injury from projectiles and other debris
For SecureFit 600.
Documents
Specifiche
- Descrizione:Foam insert for 3M™ SecureFit™ safety spectacles 600 series