Handheld refractometer, HHR-2N
Handheld refractometer (for honey), HHR-2N, 12 − 30 % Bx, 0,1 %
Codice catalogo 635-0140
Nome del fornitore: Atago
Vai a
Product Details & Documents
This refractometer is designed to determine the water content in honey.
- Scale to measure the percentage of water
- Refractive index (RI) of sucrose
- Determination of moisture in honey
Specifically suitable for use in food applications.
Specifiche
- Modello:HHR-2N
- Range di misura:12 − 30 % Bx
- Risoluzione:0,1 %
- Descrizione:Handheld refractometer (for honey)