VWR®, Turbidity standards Set
Set of turbidity standards TSK (< 0,1; 20; 200; 800 NTU), 200 NTU, 4 sealed cells, 1 KIT
Nome del fornitore: VWR Chemicals
VWR®, Turbidity standards Set
Codice catalogo 84209.600
Nome del fornitore: VWR Chemicals
Numero CAS:
Vai a
Product Details & Documents
Turbidity standards in kit form or alternatively as formazine stock solutions which allows users to create their own defined standards.
- Set of standards designed for use with turbidity meters TIR 200 and TIR 210 with a measuring range from 0,01 to 1100 NTU
- Ideal for units that measure scattered light at an angle of 90 0, as stipulated in EN ISO 7027
Certifications: ISO 7027
Informazioni per l'ordine: Kit supplied in handy case
Documents
Certificates
Cerca per numero di lotto84209_600
Immagini 360
SDS_IT_CH_VWRC84209 SDS_IT_CH_VWRC84209.pdf sds_VWRC84209
Scheda di sicurezza
VWR® for Calibrations
Letteratura sul prodotto
Specifiche
Specifiche
- Tipo di conf.:4 sealed cells
- Conf:1 KIT
- Torbidità:200 NTU
- Descrizione:Set of turbidity standards TSK (< 0,1; 20; 200; 800 NTU)