Reusable earplugs, Twisters® Cord
Catalog # MOLD644101
Supplier: Moldex
Specifications
- Colore:verde
- Descrizione:Reusable earplugs with cord
- Conf:80 PAIR
Specifications
About this item
Twisters® earplugs have a soft foam pod for a comfortable, snug seal.
- Large handle for easy insertion and removal
- No rolling necessary – plugs stay hygienic clean and can be used more than once
- Easy in-ear fit adjustment due to unique multi-curved stem
- Cord allows the plugs to be worn around the neck
- 100% PVC-free
Protection for applications with a high noise level that require constant hearing protection
SNR: 34 dB
Certifications: EN 352-2:2002
Ordering information: Twisters Ear plugs come with a cord which can be attached at any desired length.