VWR® Hitachi, Spare parts for HPLC system, Chromaster
HITA893-0809
- Descrizione:Injection valve seal, PEEK, for Chromaster 5210 and 5260 autosamplers
- Seq:575
- Conf:1
This spare part is suitable for Chromaster 5210 and 5260 autosamplers. For Chromaster 5210 AS from Serial No. 1309-001 and 1309T-001 onward. For Chromaster 5260 AS from Serial No. 1205-001 and 1205T-001 onward.