Box for Slides or Microplates
Per 30 vetrini o 5 piastre, traslucido, 120×41×85 mm
Codice catalogo GLWAK30
Nome del fornitore: GLW
Vai a
Product Details & Documents
This slide box with 30 compartments and a fixed blade can provide a secure hold for slides or microplates.
- Standard slides or microtitre plates (MTP)
- Can be used into freezers down to –90 °C
- Lid and safety seals can prevent the unauthorised access
It is suitable for storing, transporting, freezing as well as autoclaving the samples.
Delivery information: Supplied with snap lock lid and safety seals.
Specifiche
- Esterno L×P×A:120×41×85 mm
- Colore:traslucido
- Descrizione:Per 30 vetrini o 5 piastre
- Seq:007