Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 SFr.
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ghiere
Tappi per bottiglie
Tamper evident caps for allround bottles
Tamper evident caps for allround bottles
Tamper evident caps for allround bottles
Catalog # MUEL14.016.56
Supplier:
MUELLER KREMPEL
Expand image
Tamper evident caps for allround bottles
Catalog # MUEL14.016.56
Supplier:
MUELLER KREMPEL
Specifications
Descrizione:
Tamper evident closure with mounted casting ring for openings according to DIN-18
Conf:
100
Specifications
About this item
Product Details
Product Details